missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ISG15 Activating Enzyme/UBE1L Antibody (SR4330), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 316.00 - € 382.00
Specifications
| Antigen | ISG15 Activating Enzyme/UBE1L |
|---|---|
| Clone | SR4330 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 |
| Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041666
|
Novus Biologicals™
NBP3-27849-50ul |
50 μL |
€ 316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041675
|
Novus Biologicals™
NBP3-27849-100ul |
100 μL |
€ 382.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ISG15 Activating Enzyme/UBE1L Monoclonal antibody specifically detects ISG15 Activating Enzyme/UBE1L in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western BlotSpecifications
| ISG15 Activating Enzyme/UBE1L | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 7318 | |
| IgG | |
| Affinity purified |
| SR4330 | |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| D8UBA1, ubiquitin-activating enzyme E1 homolog B, UBA1B, UBA7, ubiquitin-activating enzyme E1, UBE1L, UBE2MGC12713, Ubiquitin-activating enzyme 7, Ubiquitin-activating enzyme E1 homolog, ubiquitin-activating enzyme E1-like, ubiquitin-activating enzyme E1-related protein, ubiquitin-activating enzyme-2, ubiquitin-like modifier activating enzyme 7, ubiquitin-like modifier-activating enzyme 7 | |
| A synthesized peptide derived from human ISG15 Activating Enzyme/UBE1L (Uniprot #: P41226) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title