Learn More
Islet-1 Antibody (CL11651) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15121B
Product Code. 30291552
  • € 737.00 / 0.10mL
Description
Islet-1 Monoclonal antibody specifically detects Islet-1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Islet-1
Monoclonal
Biotin
insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1
This antibody was developed using a synthetic peptide derived from P61371, with the exact immunogen sequence remaining proprietary.
0.1 mL
Diabetes Research, Lipid and Metabolism, Stem Cell Markers
3670
Store at 4⪚C in the dark.
IgG2a
Immunocytochemistry/Immunofluorescence
CL11651
PBS
Mouse
Protein A purified