missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ISLR2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547633
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human ISLR-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
| ISLR2 | |
| Polyclonal | |
| Unconjugated | |
| Islr2 | |
| B930052A04Rik; immunoglobulin superfamily containing leucine rich repeat 2; immunoglobulin superfamily containing leucine-rich repeat 2; immunoglobulin superfamily containing leucine-rich repeat protein 2; immunoglobulin superfamily containing leucine-rich repeat protein 2-like; Islr2; Kiaa1465; leucine-rich repeat domain and immunoglobulin domain containing axon extension protein; leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein; Linx; LOC100910729; Mbu-3; mKIAA1465; UNQ1885/PRO4329 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 57611 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q6UXK2 | |
| Islr2 | |
| Mouse myeloma cell line NS0-derived recombinant human ISLR-2 Cys20-Leu590. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction