missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISM2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00145501-B01P
This item is not returnable.
View return policy
Description
ISM2 Polyclonal antibody specifically detects ISM2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ISM2 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 145501 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| DKFZp686E0215, FLJ32147, isthmin 2 homolog (zebrafish), PSEC0137, thrombospondin and AMOP domain-containing isthmin-like protein 1, thrombospondin type-1 domain-containing protein 3, thrombospondin, type I domain-containing 3, THSD3, type I, domain containing 3 | |
| ISM2 (NP_954874.1, 1 a.a. - 483 a.a.) full-length human protein. MTPGNATPPRTPEVTPLRLELQKLPGLANTTLSTPNPDTQASASPDPRPLREEEEARLLPRTHLQAELHQHGCWTVTEPAALTPGNATPPRTQEVTPLLLELQKLPELVHATLSTPNPDNQVTIKVVEDPQAEVSIDLLAEPSNPPPQDTLSWLPALWSFLWGDYKGEEKDRAPGEKGEEKEEDEDYPSEDIEGEDQEDKEEDEEEQALWFNGTTDNWDQGWLAPGDWVFKDSVSYDYEPQKEWSPWSPCSGNCSTGKQQRTRPCGYGCTATETRTCDLPSCPGTEDKDTLGLPSEEWKLLARNATDMHDQDVDSCEKWLNCKSDFLIKYLSQMLRDLPSCPCAYPLEAMDSPVSLQDEHQGRSFRWRDASGPRERLDIYQPTARFCLRSMLSGESSTLAAQHCCYDEDSRLLTRGKGAGMPNLISTDFSPKLHFKFDTTPWILCKGDWSRLHAVLPPNNGRACTDNPLEEEYLAQLQEAKEY | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction