Learn More
Ismatec™ 2-Stop E-Lab Tubing
Product Code. 15136506
Click to view available options
Diameter (Metric) Inner:
1.42 mm
Unit Size:
Pack of 12
Product Code. 15136506
Brand: Ismatec™ 9646034
Suitable for applications using acetone, methyl-ethyl-ketone (MEK) and other highly aggressive chemicals
  • Designed for optimal operation in your Ismatec pumps.
  • Long service life.
  • Outstanding fatigue resistance due to flexing.
  • Resistant to ozone and UV light.
  • Won't crack, swell or harden.
  • Translucent yellow color.
Specifications
Color Code Yellow/Yellow
Description 2-Stop, E-Lab Tubing
Diameter (Metric) Inner 1.42 mm
Material Tygon E-Lab
Overall Length (English) 16 in.
Overall Length (Metric) 406 mm
Packaging Quantity 12/Pk.