missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISO 15009 Aromatic Hydrocarbon Internal Standard Mixture 460 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Ethylbenzene-d10∣Toluene D8 |
| CAS | 25837-05-2, 2037-26-5 |
| Analyte or Component Names | Ethylbenzene-d10∣Toluene D8 |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Industry Type | Food Testing |
| Quantity | 5 mL |
| Concentration or Composition (by Analyte or Components) | Ethylbenzene-d10 2000 μg/mL∣Toluene D8 2000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?