Learn More
Iso Promethazine Hydrochloride, TRC
Product Code. 30008583
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008583 1 g 1g
17973545 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008583 Supplier Toronto Research Chemicals Supplier No. TRCI8225001G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5568-90-1
Molecular Formula C17 H20 N2 S . Cl H
Molecular Weight (g/mol) 320.88
Synonym (2RS)-N,N-Dimethyl-2-(10H-phenothiazin-10-yl)propan-1-amine Hydrochloride, Promethazine Hydrochloride Imp. B (EP) as Hydrochloride, Isopromethazine Hydrochloride, 10H-Phenothiazine-10-ethanamine, N,N,β-trimethyl-, hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,N,β-trimethyl-, monohydrochloride (9CI), Phenothiazine, 10-[2-(dimethylamino)-1-methylethyl]-, monohydrochloride (8CI)
IUPAC Name N,N-dimethyl-2-phenothiazin-10-ylpropan-1-amine;hydrochloride
SMILES Cl.CC(CN(C)C)N1c2ccccc2Sc3ccccc13
Specifications
Quantity 1 g
InChI Formula InChI=1S/C17H20N2S.ClH/c1-13(12-18(2)3)19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H
Formula Weight 320.1114
Percent Purity >95%
Chemical Name or Material Isopromethazine Hydrochloride
Grade HPLC