Learn More
Brady™ ISO Safety Sign - Disinfect Surface, Dutch
Product Code. 30280523
Click to view available options
Dimensions (L x W):
150 x 50 mm
148 x 210 mm
210 x 297 mm
297 x 105 mm
297 x 420 mm
450 x 150 mm
600 x 200 mm
Material:
Aluminum
Laminated Polyester
Polypropylene
Unit Size:
Each
Product Code. 30280523
Brand: Brady™ 323309
Only null left
Add to basket
Add to basket
Sign to signify that the surface must be disinfected. Text is in Dutch.
  • Sign compliant to ISO 7010
Specifications
Certifications/Compliance ISO 7010, Compliant with Directive 2011/65/EU and 2015/863
Alternate Part Number M/M062/NL540/PE-450x150-1
Finish Gloss
Grade M062
Material Laminated Polyester
Quantity 1 Ea.
Mounting Type Surface Mount, Wall Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 5.91 in.
Height (Metric) 150 mm
Thickness 0.1 mm
Width (English) 17.72 in.
Width (Metric) 450 mm
Temperature Range (Metric) -40°C to +110°C
Shape Rectangular
Temperature Range (English) -40°F to +230°F
Languages Dutch
Dimensions (L x W) 450 x 150 mm
Displayed Text Oppervlak desinfecteren
Product Type Personal Hygiene Sign
Color White on Blue