Learn More
Brady™ ISO Safety Sign - Disinfect Surface, Pictogram
Product Code. p-200109845
Click to view available options
Material:
Aluminum
Laminated Polyester
Polypropylene
Diameter (Metric):
50 mm
100 mm
150 mm
200 mm
315 mm
400 mm
Unit Size:
Each
Pack of 2
Product Code. 30280432
Brand: Brady™ 323235
Only null left
Add to basket
Add to basket
Sign to signify that the surface must be disinfected. Universal pictogram with no text.
  • Sign compliant to ISO 7010
Specifications
Certifications/Compliance ISO 7010; Compliant with Directive 2011/65/EU and 2015/863
Alternate Part Number M/M062/NT/PE-DIA50-2
Finish Gloss
Grade M062
Material Laminated Polyester
Quantity 2/Pk.
Diameter (English) 1.97 in.
Diameter (Metric) 50 mm
Mounting Type Surface Mount, Wall Mount
For Use With (Application) Facility Identification, Safety Identification
Thickness 0.1 mm
Temperature Range (Metric) -40°C to +110°C
Shape Round
Temperature Range (English) -40°F to +230°F
Languages Universal
Product Type Personal Hygiene Sign
Color White on Blue