Learn More
Brady™ ISO Safety Sign Roll without Text - Warning: Radioactive Material or Ionizing Radiation
Safety sign for hazard identification.
€ 82.15 - € 139.00
Specifications
Finish | Gloss |
---|---|
Material | Laminated Polyester |
Adhesive Type | Permanent Acrylic |
Shape | Triangular |
Color | Black on Yellow |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
15500633
|
Brady™
826760 |
43 x 50 mm |
€ 139.00
Pack of 250 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15590623
|
Brady™
826758 |
22 x 23 mm |
€ 82.15
Pack of 250 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
Gloss | |
Permanent Acrylic | |
Black on Yellow |
Laminated Polyester | |
Triangular |