Learn More
Brady™ ISO Safety Sign without Text - Warning: Biological Hazard
Product Code. 15851427
Click to view available options
Dimensions (L x W):
9 x 10 mm
17 x 22 mm
22 x 25 mm
Unit Size:
Each
Pack of 13
Pack of 15
Product Code. 15851427
Brand: Brady™ 827647
Only null left
Add to basket
Add to basket
Safety sign for hazard identification.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number PIC W009-Tri 025-PE-SHEET/13
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Triangular
Dimensions (L x W) 17 x 22 mm
Color Black on Yellow