Learn More
Brady™ ISO Safety Sign without Text - Warning: Electricity
Product Code. 15537165 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
22 x 25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15537165 22 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15537165 Supplier Brady™ Supplier No. 828091
  • ONLINE EXCLUSIVE PRICE
    € 9.00
    € 8.12 / Pack of 1
    Save € 0.88
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Safety sign for hazard identification.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number PIC W012-Tri 025-PE-SHEET/13
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent Acrylic
Shape Triangular
Dimensions (L x W) 22 x 25 mm
Color Black on Yellow