missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iso-Sensitest™ Agar with Horse Blood and 20mg/L NAD

Product Code. 12958827
Click to view available options
Description:
Iso-Sensitest Agar with Horse Blood and 20mg/L NAD
Fill Volume:
19 mL
Packaging Type:
1 x 10 Plates
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12958827

Brand: Thermo Scientific™ PB0378A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless antimicrobial susceptibility tests (AST) using a specifically designed semi-defined medium.

Perform effortless antimicrobial susceptibility tests (AST) using Thermo Scientific™ Iso-Sensitest™ Agar with Horse Blood and 20mg/L NAD. Specifically designed for AST, Iso-Sensitest Agar allows the growth of the majority of microorganisms without further supplementation and has stability in mineral content and the ability to produce optimum antimicrobial zones of inhibition.

The formulation of Iso-Sensitest Agar with Horse Blood and 20mg/L NAD is carefully constructed to give a reproducible, semi-defined medium in which the undefined components are kept to a minimal level.

The amino-nitrogen base of acid-hydrolysed casein and special peptones has been supplemented with defined growth factors. Careful preparation of the nutrients ensures that trimethoprim and sulphonamide antagonists are at very low levels.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 10°C
Description Iso-Sensitest Agar with Horse Blood and 20mg/L NAD
Product Type Agar
Fill Volume 19 mL
Packaging Type 1 x 10 Plates

For In Vitro Diagnostic Use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.