missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoanastrozole Diamide, TRC
Specifications
Specifications
| Chemical Name or Material | Isoanastrozole Diamide |
| Analyte or Component Names | Isoanastrozole Diamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H23 N5 O2 |
| InChI Formula | InChI=1S/C17H23N5O2/c1-16(2,14(18)23)12-5-11(8-22-9-20-21-10-22)6-13(7-12)17(3,4)15(19)24/h5-7,9-10H,8H2,1-4H3,(H2,18,23)(H2,19,24) |
| Solution Type | Neat |
| SMILES | CC(C)(C(=O)N)c1cc(Cn2cnnc2)cc(c1)C(C)(C)C(=O)N |
| IUPAC Name | 2-[3-(1-amino-2-methyl-1-oxopropan-2-yl)-5-(1,2,4-triazol-4-ylmethyl)phenyl]-2-methylpropanamide |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 329.397 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?