missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISOC2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97495AF488
This item is not returnable.
View return policy
Description
ISOC2 Polyclonal specifically detects ISOC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ISOC2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ18582, FLJ23469, isochorismatase domain containing 2, isochorismatase domain-containing protein 2, mitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human ISOC2 fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 79763 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction