missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISOC2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079763-B01P
This item is not returnable.
View return policy
Description
ISOC2 Polyclonal antibody specifically detects ISOC2 in Human samples. It is validated for Western Blot
Specifications
| ISOC2 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH17344 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| FLJ18582, FLJ23469, isochorismatase domain containing 2, isochorismatase domain-containing protein 2, mitochondrial | |
| ISOC2 (AAH17344, 1 a.a. - 205 a.a.) full-length human protein. MAAARPSLGRVLPGSSVLFLCDMQEKFRHNIAYFPQIVSVAARMLKVARLLEVPVMLTEQYPQGLGPTVPELGTEGLRPLAKTCFSMVPALQQELDSRPQLRSVLLCGIEAQACILNTTLDLLDRGLQVHVVVDACSSRSQVDRLVALARMRQSGAFLSTSEGLILQLVGDAVHPQFKEIQKLIKEPAPDSGLLGLFQGQNSLLH | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 79763 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction