missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isocitrate Dehydrogenase 1/IDH1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32150C
This item is not returnable.
View return policy
Description
Isocitrate Dehydrogenase 1/IDH1 Polyclonal antibody specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Isocitrate Dehydrogenase 1/IDH1 | |
| Polyclonal | |
| Western Blot | |
| Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD | |
| The immunogen this antibody was made to, maps to a region between residue 364 to 414 of human Isocitrate Dehydrogenase 1 using the numbering given in entry NP_005887.2 (GeneID 3417). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3417 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction