missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isocitrate Dehydrogenase 1/IDH1 Antibody (OTI3G9), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70989JF549
This item is not returnable.
View return policy
Description
Isocitrate Dehydrogenase 1/IDH1 Monoclonal specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| Isocitrate Dehydrogenase 1/IDH1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IDH1 | |
| Synthetic peptide around the R132H mutation region of the human Isocitrate Dehydrogenase 1/IDH1 conjugated to KLH. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoassay | |
| OTI3G9 | |
| Western Blot, Immunohistochemistry, Knockout Validated | |
| Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 3417.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction