Learn More
IsoGel™ Agarose
Product Code. 11577985
Change view
Click to view available options
Quantity:
25 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11577985 25 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11577985 Supplier Lonza Supplier No. 50202
  • For isoelectric focusing (IEF)
  • No measurable EEO
  • Sufficiently rigid to be cast in vertical tubes
  • Gels cast on GelBond™ Film dry down quickly and easily to transparent permanent records
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cambrex Iso-Gel™ Agarose
Quantity 25 g