missing translation for 'onlineSavingsMsg'
Learn More

Isopeptidase T/USP5 Antibody (6G1) [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30519059 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519059 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519059 Supplier Novus Biologicals Supplier No. NBP342952MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Isopeptidase T/USP5 Monoclonal antibody specifically detects Isopeptidase T/USP5 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Isopeptidase T/USP5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6G1
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 5, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T, IsoT, ISOTubiquitin carboxyl-terminal hydrolase 5, ubiquitin isopeptidase T, ubiquitin specific peptidase 5 (isopeptidase T), ubiquitin specific protease 5 (isopeptidase T), ubiquitin thioesterase 5, Ubiquitin thiolesterase 5, ubiquitin-specific protease-5 (ubiquitin isopeptidase T), Ubiquitin-specific-processing protease 5
Host Species Mouse
Immunogen ubiquitin specific peptidase 5(isopeptidase T) - (Uniprot# P45974)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8078
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.