missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopeptidase T/USP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83089
This item is not returnable.
View return policy
Description
Isopeptidase T/USP5 Polyclonal antibody specifically detects Isopeptidase T/USP5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Isopeptidase T/USP5 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Deubiquitinating enzyme 5, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T, IsoT, ISOTubiquitin carboxyl-terminal hydrolase 5, ubiquitin isopeptidase T, ubiquitin specific peptidase 5 (isopeptidase T), ubiquitin specific protease 5 (isopeptidase T), ubiquitin thioesterase 5, Ubiquitin thiolesterase 5, ubiquitin-specific protease-5 (ubiquitin isopeptidase T), Ubiquitin-specific-processing protease 5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YVDKLEKIFQNAPTDPTQDFSTQVAKLGHGLLSGEYSKPVPESGDGERVPEQKEVQDGIAPRMFKALIGKGHPEFSTNRQQDAQEFFLHLINMVERNCRSSENPNEVF | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8078 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction