missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopeptidase T/USP5 Antibody (OTI2E3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71355H
This item is not returnable.
View return policy
Description
Isopeptidase T/USP5 Monoclonal specifically detects Isopeptidase T/USP5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Isopeptidase T/USP5 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| USP5 | |
| Full length recombinant protein of human Isopeptidase T/USP5 (NP_003472) produced in HEK293T cells. | |
| 0.1 mL | |
| 8078.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2E3 | |
| Western Blot, Immunohistochemistry | |
| Deubiquitinating enzyme 5, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T, IsoT, ISOTubiquitin carboxyl-terminal hydrolase 5, ubiquitin isopeptidase T, ubiquitin specific peptidase 5 (isopeptidase T), ubiquitin specific protease 5 (isopeptidase T), ubiquitin thioesterase 5, Ubiquitin thiolesterase 5, ubiquitin-specific protease-5 (ubiquitin isopeptidase T), Ubiquitin-specific-processing protease 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction