missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISYNA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 572.00
Specifications
| Antigen | ISYNA1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632218
|
Novus Biologicals
NBP2-48513-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656137
|
Novus Biologicals
NBP2-48513 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ISYNA1 Polyclonal antibody specifically detects ISYNA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ISYNA1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.2), 40% Glycerol | |
| 51477 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 5.5.1.4, hINO1, hIPS, INO1, INOS, inositol-3-phosphate synthase 1, IPSIPS 1, MI-1-P synthase, MIP synthase, Myo-inositol 1-phosphate synthase A1, Myo-inositol-1-phosphate synthase | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VYIPEFIAANQSARADNLIPGSRAQQLEQIRRDIRDFRSSAGLDKVIVLWTANTERFCEVIPGLNDTAENLLRTIELGLEVSPSTLFAVASILEGCAFLNGSPQNTLVP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title