Learn More
IT1t dihydrochloride, MedChemExpress
Product Code. 16492736
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492736 25 mg 25mg
16454616 100 mg 100mg
16443378 10 mg 10mg
16429906 50 mg 50mg
16406107 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16492736 Supplier Medchem Express Supplier No. HY101458A
MedChemExpress IT1t dihydrochloride is a potent CXCR4 antagonist; inhibits CXCL12/CXCR4 interaction with an IC50 of 2.1 nM.
IT1t dihydrochloride is a potent CXCR4 antagonist
Specifications
Chemical Name or Material IT1t dihydrochloride
Physical Form Solid
CAS 1092776-63-0
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H36Cl2N4S2
For Use With (Application) COVID-19-anti-virus
Solubility Information H2O : 50 mg/mL (104.26 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (62.56 mM)
SMILES CC1(C)CN2C(CS/C(NC3CCCCC3)=N\C4CCCCC4)=CSC2=N1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 479.57
Formula Weight 479.57
Percent Purity 99.89%
Grade Research
Show More Show Less
Research purposes only