missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITFG1 Antibody (936213), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB89001MFV450
This item is not returnable.
View return policy
Description
ITFG1 Monoclonal antibody specifically detects ITFG1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| ITFG1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 936213 | |
| Flow Cytometry, CyTOF-ready | |
| CDA08, integrin alpha FG-GAP repeat containing 1,2310047C21Rik, Integrin-alpha FG-GAP repeat-containing protein 1, Protein TIP, T-cell immunomodulatory protein, TIPT cell immunomodulatory protein | |
| Human embryonic kidney cell line HEK293-derived human ITFG1, Met1-Ile566, Accession # Q8TB96 | |
| RUO | |
| 81533 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction