missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITFG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 156.00 - € 476.00
Specifications
| Antigen | ITFG1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17770614
![]() |
Invitrogen™
PA5119209 |
200 μL |
€ 476.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17773214
|
Invitrogen™
PA5121819 |
20 μL |
€ 156.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITFG1 Polyclonal Antibody for Western Blot, IHC (P)
Modulator of T-cell function. Has a protective effect in graft versus host disease model.Specifications
| ITFG1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Itfg1 | |
| 2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP | |
| Itfg1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8TB96, Q99KW9 | |
| 71927, 81533 | |
| Recombinant protein Integrin Alpha FG GAP Repeat Containing Protein 1. The antigen corresponds to amino acid range 55-306 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title