Learn More
Invitrogen™ ITFG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590805
Description
Immunogen sequence: LWGFVPFVDE QQPTEIPIPI TLHIGDYNMD GYPDALVILK NTSGSNQQAF LLENVPCNNA SCEEARRMFK VYWELTDLNQ IKDAMVATFF DIYEDGILDI VVLSKGYTKN DFAIHTLKNN FEADAYFVKV IVLSGLCSND CPRKITPFGV NQPGPYIMYT TVDANGYLKN GSAGQLSQSA HLALQLPYNV LGLGRSANFL DHLYVGIPRP SGEKSIRKQE WTAIIPNSQL IVIPYPHNVP RSWSAKLYLT PSNIVLLTAI ALIGVCVFIL AIIGILHWQE KKADDREKRQ EAHRFHFDAM; Positive Samples: LO2, A-431, Mouse heart, Mouse pancreas, Mouse testis, ; Cellular Location: Membrane, Single-pass type I membrane protein.
Modulator of T-cell function. Has a protective effect in graft versus host disease model.
Specifications
| ITFG1 | |
| Polyclonal | |
| Unconjugated | |
| Itfg1 | |
| 2310047C21Rik; AI314457; CDA08; CDA08-like protein; D8Wsu49e; hypothetical protein CDA08; integrin alpha FG-GAP repeat containing 1; integrin-alpha FG-GAP repeat-containing protein 1; ITFG1; Linkin; LNKN-1; Protein TIP; T-cell immunomodulatory protein; TIP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171083, 71927, 81533 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.22 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8R4E1, Q8TB96, Q99KW9 | |
| Itfg1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.