Learn More
ITFG2 Monoclonal Antibody (OTI 2H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503044S
This item is not returnable.
View return policy
Description
ITFG2 Monoclonal antibody specifically detects ITFG2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ITFG2 | |
| Monoclonal | |
| 0.54 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14543 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9021 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI 2H5 | |
| Unconjugated | |
| Socs3 | |
| 2700050P07Rik; AI646725; integrin alpha FG-GAP repeat containing 2; integrin-alpha FG-GAP repeat-containing protein 2; integrin-alpha FG-GAP repeat-containing protein 2; KICSTOR complex protein ITFG2; ITFG2; KICSTOR complex protein ITFG2; MDS028; RGD1311944 | |
| Full length human recombinant protein of human SOCS3 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |