Learn More
ZFAND5 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800482
Encompass_Preferred
 View more versions of this product
Product Code. 17518382
  • € 540.87 / 100µL
Description
ZNF216 (ZFAND5) is a potent inhibitory factor for osteoclast differentiation and the mechanism is unlikely due to direct attenuation of the NF-kappa B pathway. It also has redundant and distinct roles in regulating apoptosis.
Specifications
Specifications
ZFAND5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O76080
ZFAND5
Full length human recombit protein of human ZFAND5 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1D8
Unconjugated
ZFAND5
ZA20D2, ZFAND5A, ZNF216
Mouse
Affinity Chromatography
RUO
7763
-20° C, Avoid Freeze/Thaw Cycles