missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITFG2 Monoclonal Antibody (OTI3A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503049S
This item is not returnable.
View return policy
Description
ITFG2 Monoclonal antibody specifically detects ITFG2 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ITFG2 | |
| Monoclonal | |
| 0.36 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q969R8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55846 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI3A6 | |
| Unconjugated | |
| ITFG2 | |
| 2700050P07Rik; AI646725; integrin alpha FG-GAP repeat containing 2; integrin-alpha FG-GAP repeat-containing protein 2; integrin-alpha FG-GAP repeat-containing protein 2; KICSTOR complex protein ITFG2; ITFG2; KICSTOR complex protein ITFG2; MDS028; RGD1311944 | |
| Full length human recombinant protein of human ITFG2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction