missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITFG3 Antibody (OTI3B3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72253H
This item is not returnable.
View return policy
Description
ITFG3 Monoclonal specifically detects ITFG3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ITFG3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ITFG3 | |
| Full length recombinant protein of human ITFG3 (NP_114428) produced in HEK293T cells. | |
| 0.1 mL | |
| 83986.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3B3 | |
| Western Blot, Immunohistochemistry | |
| C16orf9, chromosome 16 open reading frame 9, DKFZP761D0211, FLJ32603, gs19, integrin alpha FG-GAP repeat containing 3, protein ITFG3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction