missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITFG3 Monoclonal Antibody (OTI3B3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526160
This item is not returnable.
View return policy
Description
ITFG3 Monoclonal Antibody for IHC (P), Western Blot
ITFG3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cell surface; extracellular exosome; integral component of membrane.
Specifications
| ITFG3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H0X4 | |
| FAM234A | |
| Full length human recombinant protein of ITFG3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B3 | |
| Unconjugated | |
| FAM234A | |
| C16orf9; Fam234a; family with sequence similarity 234 member A; family with sequence similarity 234, member A; gs19; integrin alpha FG-GAP repeat containing 3; ITFG3; Protein FAM234A; protein ITFG3; RGD1311484 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 83986 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction