missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGA4 Monoclonal Antibody (R1-2), PE
Rat Monoclonal Antibody
€ 220.00 - € 506.00
Specifications
| Antigen | ITGA4 |
|---|---|
| Clone | R1-2 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, do not freeze |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15374081
|
Invitrogen™
MA517808 |
50 μg |
€ 220.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15384081
|
Invitrogen™
MA517809 |
300 μg |
€ 506.00
300µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITGA4 Monoclonal Antibody for Flow
The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.Specifications
| ITGA4 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| PE | |
| Rat | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| Q00651 | |
| 16401 | |
| Peyers Patch HEV binding lymphoma line (TK1). | |
| Antibody |
| R1-2 | |
| 4°C, do not freeze | |
| Monoclonal | |
| Liquid | |
| IgG2b | |
| Mouse | |
| ITGA4 | |
| 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
| ITGA4 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title