missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA5, anti-Human, Clone: M200 (Volociximab), Abnova™
Monoclonal Antibody
Brand: Abnova RAB00061.200ug
This item is not returnable.
View return policy
Specifications
| ITGA5 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| CD49e/FNRA/VLA5A | |
| Original antibody is raised against heterodimeric alpha 5 beta 1 Fc fusion protein. | |
| RUO | |
| 3678/3688 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Blocking Assay, ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| M200 (Volociximab) | |
| Blocking ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| integrin, alpha 5 (fibronectin receptor, alpha polypeptide) | |
| ITGA5 | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG4 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur