missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITGA8 Monoclonal Antibody (CL7304)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531449

Product Code. 16344894

  • € 590.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HKEEEVGPLV EHIYELHNIG PSTISDTILE VGWPFSARDE FLLYIFHIQT LGPLQCQPNP NINPQDIKPA ASPEDTPELS AFLRNSTIPH LVRKRDVHVV EFHRQSPAKI LNCTNIECLQ ISCA Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.

Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITGA8
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P53708
ITGA8
Recombinant protein fragment of Human ITGA8 (Product #RP-100997)
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
CL7304
Unconjugated
ITGA8
AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265
Mouse
Protein A
RUO
8516
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.