missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGAL, anti-Human, Clone: hu1124 (Efalizumab), Abnova™
Monoclonal Antibody
Brand: Abnova RAB00051.200ug
This item is not returnable.
View return policy
Specifications
| ITGAL | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| CD11A/LFA-1/LFA1A | |
| Original antibody is raised against human ITGAL. | |
| RUO | |
| 3683 | |
| Human | |
| Antibody | |
| IgG1 κ |
| Blocking Assay, Flow Cytometry | |
| hu1124 (Efalizumab) | |
| Blocking Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide) | |
| ITGAL | |
| 200 μg | |
| Primary | |
| This antibody binds to human ITGAL. | |
| Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction