missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGAM Rat anti-Human, Mouse, PerCP-Cy5.5, Clone: M1/70, Abnova™
Rat monoclonal antibody raised against Itgam.
Brand: Abnova MAB12217.100ug
This item is not returnable.
View return policy
Specifications
| ITGAM | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| Flow Cytometry The optimal working dilution should be determined by the end user. | |
| CD11b/CD18/CR3/CR3A/Cd11b/F730045J24Rik/Ly-40/MAC1/Mac-1/Mac-1a | |
| Rat | |
| Affinity Chromatography | |
| RUO | |
| 16409 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry | |
| M1/70 | |
| Rat monoclonal antibody raised against Itgam. | |
| In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
| Itgam | |
| Itgam | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction