missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB3 Mouse anti-Human, Clone: Y2/51, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14568.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human ITGB3.
- High specificity
- High reactivity
Specifications
| ITGB3 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL) The optimal working d | |
| P05106 | |
| ITGB3 | |
| Phytohemagglutinin (PHA)-stimulated peripheral blood cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
| Y2/51 | |
| Mouse monoclonal antibody raised against native human ITGB3. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| CD61/GP3A/GPIIIa | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3690 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction