missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB3BP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544255
This item is not returnable.
View return policy
Description
Peptide sequence: TGTCQMSLFA SPTSSEEQKH RNGLSNEKRK KLNHPSLTES KESTTKDNDE Sequence homology: Human: 100%.
ITGB3BP is a transcription coregulator that can have both coactivator and corepressor functions. Isoform 1, but not other isoforms, is involved in the coactivation of nuclear receptors for retinoid X (RXRs) and thyroid hormone (TRs) in a ligand-dependent fashion. ITGB3BP acts as a transcriptional corepressor via its interaction with the NFKB1 NF-kappa-B subunit, possibly by interfering with the transactivation domain of NFKB1. It induces apoptosis in breast cancer cells, but not in other cancer cells, via a caspase-2 mediated pathway that involves mitochondrial membrane permeabilization but does not require other caspases. ITGB3BP may also act as an inhibitor of cyclin A-associated kinase. ITGB3BP may be involved in incorporation of newly synthesized CENPA into centromeres via its interaction with the CENPA-NAC complex.
Specifications
| ITGB3BP | |
| Polyclonal | |
| Unconjugated | |
| ITGB3BP | |
| 4930471O16Rik; AU022583; beta 3 endonexin; beta-3-endonexin; beta3-endonexin; Cenpr; CENP-R; Centromere protein R; HSU37139; integrin beta 3 binding protein (beta3-endonexin); integrin beta-3-binding protein; integrin subunit beta 3 binding protein; Itgb3bp; Nrif3; Nuclear receptor-interacting factor 3; TAP20 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23421 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13352 | |
| ITGB3BP | |
| Synthetic peptide directed towards the N-terminal of human ITGB3BP (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction