missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576390
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ITGB8 (Integrin beta 8) is a member of the integrin beta chain family and is a single-pass type I membrane protein with a VWFA domain and four cysteine-rich repeats. This protein noncovalently binds to an alpha subunit to form a heterodimeric integrin complex. In general, integrin complexes mediate cell-cell and cell-extracellular matrix interactions and this complex plays a role in human airway epithelial proliferation. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| ITGB8 | |
| Polyclonal | |
| Unconjugated | |
| ITGB8 | |
| 4832412O06Rik; integrin beta 8; Integrin beta8; integrin beta-8; integrin subunit beta 8; integrin, beta 8; ITGB8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 320910, 362800, 3696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26012 | |
| ITGB8 | |
| Recombinant fusion protein of human ITGB8. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction