missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH3 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97601APC
This item is not returnable.
View return policy
Description
ITIH3 Polyclonal antibody specifically detects ITIH3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ITIH3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP | |
| Produced in rabbits immunized with E. coli-derived Human ITIH3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3699 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction