missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITIH3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522232
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, BCL-1. Predicted reactivity: Mouse (87%), Rat (87%), Pig (85%), Rabbit (91%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the heavy chain subunit of the pre-alpha-trypsin inhibitor complex. This complex may stabilize the extracellular matrix through its ability to bind hyaluronic acid. Polymorphisms of this gene may be associated with increased risk for schizophrenia and major depressive disorder. This gene is present in an inter-alpha-trypsin inhibitor family gene cluster on chromosome 3.
Specifications
| ITIH3 | |
| Polyclonal | |
| Unconjugated | |
| ITIH3 | |
| AW108094; H3P; inter-alpha (globulin) inhibitor H3; inter-alpha (globulin) inhibitor, H3 polypeptide; inter-alpha trypsin inhibitor, heavy chain 3; inter-alpha-inhibitor H3 chain; inter-alpha-inhibitor heavy chain 3; inter-alpha-trypsin inhibitor heavy chain 3; inter-alpha-trypsin inhibitor heavy chain H3; Intin3; ITI heavy chain H3; ITIH3; Itih-3; ITI-HC3; PAIHC3; pre-alpha (globulin) inhibitor, H3 polypeptide; pre-alpha-inhibitor heavy chain 3; pre-alpha-inhibitor, heavy chain 3; serum-derived hyaluronan-associated protein; SHAP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16426, 3699 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q06033, Q61704 | |
| ITIH3 | |
| Recombinant protein encompassing a sequence within the center region of human ITIH3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction