missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH3 Rabbit anti-Human, Clone: 49, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00
Specifications
| Antigen | ITIH3 |
|---|---|
| Clone | 49 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233508
|
Novus Biologicals
NBP2-90329-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237027
|
Novus Biologicals
NBP2-90329-100UL |
100 μg |
N/A
|
N/A | |||||
Description
ITIH3 Monoclonal specifically detects ITIH3 in Human samples. It is validated for ELISA.Specifications
| ITIH3 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3699 | |
| IgG | |
| Protein A purified |
| 49 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP | |
| Recombinant Human ITIH3 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title