missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | ITK |
|---|---|
| Dilution | Western Blot 1:2000 - 1:6000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229149
|
Novus Biologicals
NBP3-33227-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229526
|
Novus Biologicals
NBP3-33227-20ul |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITK Monoclonal antibody specifically detects ITK in Human samples. It is validated for ELISA,Western BlotSpecifications
| ITK | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 3702 | |
| IgG | |
| Affinity purified |
| Western Blot 1:2000 - 1:6000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk | |
| Recombinant protein of human ITK. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title