missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ITK |
|---|---|
| Clone | 1F9D8 |
| Concentration | 1624 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235765
|
Proteintech
67703-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234791
|
Proteintech
67703-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ITK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ITK. This antibody recognizes human antigen. The ITK antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| ITK | |
| 1624 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3702 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. |
| 1F9D8 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| EMT; LYK; MGC126257; MGC126258; PSCTK2 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title