missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITM2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99451-50ul
This item is not returnable.
View return policy
Description
ITM2A Polyclonal antibody specifically detects ITM2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ITM2A | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin 1:500-1:5000 | |
| BRICHOS domain containing 2A, E25ABRICD2A, integral membrane protein 2A, Protein E25 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ITM2A. | |
| 50 μg | |
| Stem Cell Markers | |
| 9452 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction