missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITM2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100581
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ITM2A.
This gene encodes a type II membrane protein that belongs to the ITM2 family. Studies in mouse suggest that it may be involved in osteo- and chondrogenic differentiation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ITM2A | |
| Polyclonal | |
| Unconjugated | |
| ITM2A | |
| AW108051; BRICD2A; BRICHOS domain containing 2A; E25; E25A; Integral membrane protein 2A; Itm2; ITM2A; Protein E25; UNQ603/PRO1189 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16431, 317218, 9452 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43736, Q61500 | |
| ITM2A | |
| A synthesized peptide derived from human ITM2A(Accession O43736), corresponding to amino acid residues C73-I123. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction