missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITM2B Antibody (OTI1C11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71460UV
This item is not returnable.
View return policy
Description
ITM2B Monoclonal specifically detects ITM2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| ITM2B | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ITM2B | |
| Full length recombinant protein of human ITM2B (NP_068839) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1C11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| ABRI, ABri/ADan amyloid peptide, BRI2, BRICD2B, BRICHOS domain containing 2B, BRIFBD, E25B, E3-16, integral membrane protein 2B, Protein E25B, Transmembrane protein BRI | |
| Mouse | |
| Affinity Purified | |
| Neuroscience, Signal Transduction | |
| 9445.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction