missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPK1 Antibody (OTI1F5), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71052DL594
This item is not returnable.
View return policy
Description
ITPK1 Monoclonal antibody specifically detects ITPK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ITPK1 | |
| Monoclonal | |
| DyLight 594 | |
| EC 2.7.1.134, EC 2.7.1.159, inositol 13,4-triphosphate 5/6 kinase, inositol 13,4-trisphosphate 5/6 kinase, Inositol 13,4-trisphosphate 5/6-kinase, inositol-tetrakisphosphate 1-kinase, Inositol-triphosphate 5/6-kinase, Ins(13,4)P(3) 5/6-kinase, ITRPK1 | |
| Recombinant protein fragment corresponding to amino acids 309-391 of human ITPK1 (NP_001136065) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 3705 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1F5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction