missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPR1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Description
Sequence: A synthetic peptide corresponding to amino acids 1565-1615 of human ITPR1.
Specifications
Specifications
| Antigen | ITPR1 |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. |
| Formulation | In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gene | inositol 1,4,5-triphosphate receptor, type 1 |
| Gene Alias | INSP3R1/IP3R/IP3R1/SCA15/SCA16 |
| Gene Symbols | ITPR1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?